Please login to your online account to display your discounted pricing

Phenol Red, 0.1%, Aqueous, Certified, LabChem™

$63.86 - $63.86

Chemical Identifiers

Chemical Name or Material Phenol Red
CAS 7732-18-5
CAS Max % 99.9
CAS 34487-61-1
CAS Max % 0.1
Name Note 0.1%, Aqueous
Purity Grade Certified Grade
Linear Formula C19H13O5SNa
Formula Weight 376.36
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC182701

 
lab chem
LC182701
500mL Each for $63.86
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 376.36
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Quantity 500mL
Color Clear, orange
CAS 7732-18-5
CAS Max % 99.9
CAS 34487-61-1
Linear Formula C19H13O5SNa
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Chemical Name or Material Phenol Red
Recommended Storage Room Temperature
Name Note 0.1%, Aqueous
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Solubility Soluble in water
Packaging Poly Bottle
Identification Passes Test
Physical Form Liquid
CAS Max % 0.1
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature