Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenol, Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer


Quantity 100mL

Manufacturer: alfa aesar™  J67050AE

 View more versions of this product

Catalog No. AAJ67050AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 94.11
MDL Number MFCD00002143
Chemical Name or Material Phenol
CAS 108-95-2
TSCA TSCA
DOT Information Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: PHENOL SOLUTION
Density 1.060g/mL
UN Number UN2821
Name Note Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer
Sensitivity Light Sensitive
Physical Form Liquid
Color Clear
Quantity 100mL

Specifications

Chemical Name or Material Phenol
CAS 108-95-2
Name Note Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer
Formula Weight 94.11
MDL Number MFCD00002143
UN Number UN2821