Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenol, Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Name Note Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer
Formula Weight 94.11
MDL Number MFCD00002143
UN Number UN2821
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67050AE

  SDS
alfa aesar™
J67050AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67050AN

  SDS
alfa aesar™
J67050AN
400mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 94.11
Chemical Name or Material Phenol
MDL Number MFCD00002143
TSCA TSCA
Color Clear
Quantity 100mL
Sensitivity Light Sensitive
Name Note Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer
Physical Form Liquid
Density 1.060g/mL
CAS 108-95-2
UN Number UN2821
DOT Information Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: PHENOL SOLUTION