Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenol, Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer


Quantity 400mL

Manufacturer: alfa aesar™  J67050AN

 View more versions of this product

Catalog No. AAJ67050AN

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 94.11
CAS 108-95-2
Chemical Name or Material Phenol
TSCA TSCA
Physical Form Liquid
MDL Number MFCD00002143
UN Number UN2821
Color Clear
Quantity 400mL
Sensitivity Light Sensitive
Name Note Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer
Density 1.060g/mL
DOT Information Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: PHENOL SOLUTION

Specifications

Chemical Name or Material Phenol
CAS 108-95-2
Name Note Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer
Formula Weight 94.11
MDL Number MFCD00002143
UN Number UN2821