Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenol, Ready-to-Use, buffer saturated solution pH 4.5

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Name Note Ready-to-Use, buffer saturated solution
Formula Weight 94.11
MDL Number MFCD00002143
UN Number UN2821
Beilstein 6428463
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67158AE

  SDS
alfa aesar™
J67158AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67158AN

  SDS
alfa aesar™
J67158AN
400mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 94.11
Chemical Name or Material Phenol
CAS 108-95-2
Health Hazard 2 GHS H Statement
H311-H331-H314-H341-H373-H227-H302
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Health Hazard 1 Danger
Physical Form Liquid
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Sensitivity Light Sensitive
DOT Information Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: PHENOL SOLUTION
Quantity 100mL
pH 4.5
Beilstein 6428463
Name Note Ready-to-Use, buffer saturated solution
Density 1.06g/mL
UN Number UN2821
Color Clear
MDL Number MFCD00002143
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H331-H314-H341-H373-H227-H302
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.