Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenol on polystyrene, ca 3.5 mmol/g

Chemical Identifiers

Chemical Name or Material Phenol on polystyrene
MDL Number MFCD03703209
Synonym Poly(4-hydroxy)styrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1959403

 
alfa aesar™
L19594-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Quantity 1g
Concentration ≈3.5mmol/g
Quantity 1g
Chemical Name or Material Phenol on polystyrene
MDL Number MFCD03703209
Synonym Poly(4-hydroxy)styrene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.