Please login to your online account to display your discounted pricing

Phenol, Molecular Biology Grade, Calbiochem™, EMD Millipore

$196.01 - $196.01

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Physical Form Solid
Molecular Formula C6H6O
Formula Weight 94.1
Color Colorless
UN Number UN 1671
Melting Point 40.8°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

516724500GM

 
emd millipore corporation
516724500GM
Each for $196.01
Description & Specifications

Specifications

Purity Grade GC Grade
Formula Weight 94.1
Boiling Point 181.8°C
Melting Point 40.8°C
pH 5 (50g/L at 20°C)
Quantity 500g
Purity Grade Notes Molecular biology grade
Assay ≥99.0%
Molecular Formula C6H6O
Solubility Information Water: 84g/L at 20°C soluble
Additional Information Packaged under inert gas
Health Hazard 1 H301-H311-H331: Toxic if swallowed. In contact with skin or if inhaled
Vapor Pressure 0.2 hPa (20°C)
Density 1.06g/cm3 (20°C)
Health Hazard 2 H314: Causes severe skin burns and eye damage.
Recommended Storage -20°C (-4°F), Tightly closed. Dry. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized person.
DOT Information DOT Class: 6.1, Packaging Group: II
RTECS Number SJ3325000
Chemical Name or Material Phenol
Odor Characteristic
Physical Form Solid
CAS 108-95-2
UN Number UN 1671
Color Colorless
Ignition Point 595°C (1103°F)
Health Hazard 3 H341: Suspected of causing genetic defects.
Safety and Handling

H301-H311-H331: Toxic if swallowed. In contact with skin or if inhaled

H341: Suspected of causing genetic defects.

H314: Causes severe skin burns and eye damage.

-20°C (-4°F), Tightly closed. Dry. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized person.