Please login to your online account to display your discounted pricing

Phenol (Fused Crystals/Laboratory), Fisher Chemical

$414.77 - $441.35

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Purity Grade Laboratory Grade
Physical Form Solid
Molecular Formula C6H6O
Linear Formula C6H5OH
Formula Weight 94.11
MDL Number MFCD00002143
Synonym Carbolic Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A91I-212

 
fisher scientific company
BPA91I212
Glass Bottle 2.5kg Each for $441.35

A91I-500

 
fisher scientific company
BPA91I500
Glass Bottle 500g Each for $414.77
Description & Specifications

Specifications

Formula Weight 94.11
Melting Point 39°C
pH 4 to 5
Purity Grade Laboratory Grade
Boiling Point 182°C
Quantity 2.5kg
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Vapor Pressure 0.4mbar at 20°C
Viscosity 3.437 mPa/s (50°C)
Density 1.070g/cm³
Color Colorless
Chemical Name or Material Phenol
Identification Pass Test
Packaging Glass Bottle
Molecular Formula C6H6O
Linear Formula C6H5OH
MDL Number MFCD00002143
CAS 108-95-2
ChemAlert Storage Symbol White
Synonym Carbolic Acid
Additional Information Vapor Pressure: 0.4mbar at 20°C
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Combustible material. Causes burns by all exposure routes. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:2
Instability:1
Safety and Handling
 

Emergency Overview
Combustible material. Causes burns by all exposure routes. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:2
Instability:1

DANGER!