Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenol-d1, 95% (Isotopic)

Chemical Identifiers

Chemical Name or Material Phenol-d1
CAS 1003-66-3
Name Note Isotopic
Molecular Formula C6H5DO
Formula Weight 95.12
MDL Number MFCD01075468
UN Number UN1671
Synonym Phenol-4-d1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5110003

  SDS
alfa aesar™
H5110003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5110006

  SDS
alfa aesar™
H5110006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 95.12
Synonym Phenol-4-d1
Assay 95%
Health Hazard 2 GHS H Statement
H300-H310-H330-H314-H318-H341-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Boiling Point 182°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1671
Melting Point 40° to 42°C
MDL Number MFCD01075468
Quantity 1g
Molecular Formula C6H5DO
Name Note Isotopic
CAS 1003-66-3
Chemical Name or Material Phenol-d1
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H314-H318-H341-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.