Please login to your online account to display your discounted pricing

Phenol (Crystallized), Fisher BioReagents


Quantity 500g
Packaging Amber Glass/Poison Pack

Manufacturer: fisher scientific company  BP226500

 View more versions of this product

Catalog No. BP226-500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 94.11
pH 4 to 5
Boiling Point 182°C
Melting Point 39°C
Quantity 500g
Merck Index 15, 7354
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Density 1.070g/cm³
Color Colorless
Vapor Pressure 0.4mbar at 20°C
Viscosity 3.437 mPa/s (50°C)
Heavy Metals (as Pb) 0.01% max.
Synonym Carbolic Acid
Assay Percent Range ≥99.0 %
ChemAlert Storage Symbol White
Molecular Formula C6H6O
Additional Information Vapor Pressure: 0.4mbar at 20°C
Packaging Amber Glass/Poison Pack
Saturation with Tris Pass Test
Linear Formula C6H5OH
Identification Pass Test
Chemical Name or Material Phenol
Moisture 0.5% max.
CAS 108-95-2

Specifications

Chemical Name or Material Phenol
CAS 108-95-2
Physical Form Solid
Assay ≥99.0%
Assay Percent Range ≥99.0 %
Molecular Formula C6H6O
Linear Formula C6H5OH
Formula Weight 94.11
Synonym Carbolic Acid
Accessories
 

SDS & Safety Documents