Please login to your online account to display your discounted pricing

Phenol (Crystallized), Fisher BioReagents

$293.93 - $409.15

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Physical Form Solid
Assay ≥99.0%
Assay Percent Range ≥99.0 %
Molecular Formula C6H6O
Linear Formula C6H5OH
Formula Weight 94.11
Synonym Carbolic Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP226-100

  SDS
fisher scientific company
BP226100
Amber Glass/Poison Pack 100g Each for $293.93

BP226-500

  SDS
fisher scientific company
BP226500
Amber Glass/Poison Pack 500g Each for $409.15
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 94.11
Boiling Point 182°C
Melting Point 39°C
pH 4 to 5
Quantity 100g
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Vapor Pressure 0.4mbar at 20°C
Viscosity 3.437 mPa/s (50°C)
Density 1.070g/cm³
Color Colorless
Merck Index 15, 7354
Chemical Name or Material Phenol
Heavy Metals (as Pb) 0.01% max.
Identification Pass Test
Packaging Amber Glass/Poison Pack
Molecular Formula C6H6O
Linear Formula C6H5OH
CAS 108-95-2
ChemAlert Storage Symbol White
Synonym Carbolic Acid
Moisture 0.5% max.
Assay Percent Range ≥99.0 %
Saturation with Tris Pass Test
Additional Information Vapor Pressure: 0.4mbar at 20°C

Phenol is used in nucleic acid purification procedures.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling