Please login to your online account to display your discounted pricing

Phenol/Chloroform/Isoamyl alcohol (25:24:1), Stabilized, for Molecular Biology, DNAs, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC327111000

 View more versions of this product

Catalog No. AC327111000

  • / Each
 

Description & Specifications

Chemical Identifiers

pH 7.8 to 8.2 (25°C)
Physical Form Solution
Formula Weight 94.11
Quantity 100mL
Color Clear yellow to orange
Merck Index 14, 7241
Treatment(s) Stabilized
Purity Grade For molecular biology
Density 1.2800g/mL
UN Number 2927
CAS 136112-00-0
MDL Number MFCD00133763
DNase RNase Protease act none detected
Heavy Metals (as Pb) 1ppm max.
Health Hazard 2 GHS P Statement Suspected of causing genetic defects. Suspected of causing cancer. Causes damage to organs through prolonged or repeated exposure if inhaled. Causes severe skin burns and eye damage. Toxic if inhaled. Toxic in contact with skin. Toxic if swallowed.
Iron (Fe) 1ppm max.
CAS 67-66-3
Packaging Glass bottle
Stabilizer 0.08 to 0.12% hydroxyquinoline
CAS 123-51-3
Chemical Name or Material Phenol/Chloroform/Isoamyl alcohol (25:24:1), stabilized
Molecular Formula C6H6O
CAS 136112-00-0
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses
Name Note DNase, RNase and Protease Free

Specifications

Chemical Name or Material Phenol/Chloroform/Isoamyl alcohol (25:24:1), stabilized
Name Note DNase, RNase and Protease Free
CAS 136112-00-0
CAS 67-66-3
CAS 123-51-3
Purity Grade For molecular biology
Molecular Formula C6H6O
Formula Weight 94.11
MDL Number MFCD00133763
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses

GHS P Statement Suspected of causing genetic defects. Suspected of causing cancer. Causes damage to organs through prolonged or repeated exposure if inhaled. Causes severe skin burns and eye damage. Toxic if inhaled. Toxic in contact with skin. Toxic if swallowed.

SDS & Safety Documents