Please login to your online account to display your discounted pricing

Phenol/Chloroform/Isoamyl alcohol (25:24:1), Stabilized, for Molecular Biology, DNAs, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC327110025

 View more versions of this product

Catalog No. AC327110025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

pH 7.8 to 8.2 (25°C)
Physical Form Solution
Formula Weight 94.11
Quantity 2.5L
Color Clear yellow to orange
Treatment(s) Stabilized
Purity Grade For molecular biology
Density 1.2800g/mL
Merck Index 14, 7241
UN Number 2927
CAS 136112-00-0
Health Hazard 1 Danger
Iron (Fe) 1ppm max.
Chemical Name or Material Phenol/Chloroform/Isoamyl alcohol (25:24:1), stabilized
MDL Number MFCD00133763
DNase RNase Protease act none detected
Heavy Metals (as Pb) 1ppm max.
Health Hazard 2 GHS P Statement Suspected of causing genetic defects. Suspected of causing cancer. Causes damage to organs through prolonged or repeated exposure if inhaled. Causes severe skin burns and eye damage. Toxic if inhaled. Toxic in contact with skin. Toxic if swallowed.
CAS 67-66-3
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses
Packaging Glass bottle
Stabilizer 0.08 to 0.12% hydroxyquinoline
CAS 123-51-3
Name Note DNase, RNase and Protease Free
Molecular Formula C6H6O
CAS 136112-00-0

Specifications

Chemical Name or Material Phenol/Chloroform/Isoamyl alcohol (25:24:1), stabilized
Name Note DNase, RNase and Protease Free
CAS 136112-00-0
CAS 67-66-3
CAS 123-51-3
Purity Grade For molecular biology
Molecular Formula C6H6O
Formula Weight 94.11
MDL Number MFCD00133763
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses

GHS P Statement Suspected of causing genetic defects. Suspected of causing cancer. Causes damage to organs through prolonged or repeated exposure if inhaled. Causes severe skin burns and eye damage. Toxic if inhaled. Toxic in contact with skin. Toxic if swallowed.

SDS & Safety Documents