missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol:Chloroform:Isoamyl Alcohol, 25:24:1, OmniPur™, Calbiochem™, EMD Millipore

TE Buffered Saturated pH 6.7/8.0; DNA applications

$78.24 - $78.24

Chemical Identifiers

Assay Percent Range ≥99.0%
Chemical Name or Material Phenol:Chloroform:Isoamyl Alcohol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
6805100ML Emd Millipore Corp
6805100ML
100mL Each for $78.24
Specifications Description & Specifications

Chemical Identifiers

≥99.0%
Phenol:Chloroform:Isoamyl Alcohol

Specifications

330nm: ≤0.2AU, 405nm: ≤0.1AU, 510nm: ≤0.1AU
Molecular Biology Grade
Phenol:Chloroform:Isoamyl Alcohol
≥99.0%
100mL