Please login to your online account to display your discounted pricing

Phenol/Chloroform/Isoamyl Alcohol (25:24:1) Molecular Biology Grade, Calbiochem™, EMD Millipore

Suitable for DNA and RNA applications

$215.47 - $215.47

Chemical Identifiers

Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol (25:24:1)
UN Number 2810
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5167261SET

 
emd millipore corporation
5167261SET
Each for $215.47
Description & Specifications

Specifications

Quantity 1 set
Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol (25:24:1)
Storage Information 2°to 8°C (36°to 46°F) Tightly closed. Keep in well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons.
Assay ≥99%
UN Number 2810
Health Hazard 1 H301: Toxic if swallowed
Health Hazard 2 H312: Harmful in contact with skin
Description Supplied with 26 ml Tris-based equilibration buffer.
Health Hazard 3 H314: Causes sever skin burns and eye damage
Safety and Handling

H301: Toxic if swallowed

H314: Causes sever skin burns and eye damage

H312: Harmful in contact with skin