Please login to your online account to display your discounted pricing

Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 5.2±0.3, Liq.), Fisher BioReagents


Quantity 100mL
Packaging Coated Amber Glass/Poison Pack

Manufacturer: fisher scientific company  BP1753I100

 View more versions of this product

Catalog No. BP1753I-100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Boiling Point 96°C
Quantity 100mL
Merck Index 15, 7354
DOT Information DOT Class 6.1, : Poison
Physical Form Liquid
pH 5.2
Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol
Packaging Coated Amber Glass/Poison Pack
Recommended Storage 4°C
Identification Pass Test
Optical Density 330nm (0.2 AU max.), 405nm (0.1 AU), 510nm (0.1nm)
ChemAlert Storage Symbol White
CAS 67-66-3

Specifications

Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol
CAS 67-66-3
Physical Form Liquid
Assay ≥99.0%
Accessories
 

4°C

SDS & Safety Documents