missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents

$159.27 - $238.49

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP1754I-400 SDS View Documents Fisher BioReagents
BP1754I400
400mL Coated Amber Glass/Poison Pack Each for $238.49
BP1754I-100 SDS View Documents Fisher BioReagents
BP1754I100
100mL Coated Amber Glass/Poison Pack Each for $159.27
Description

Description

This mixture is used to purify total RNA from mixtures containing RNA, DNA, and protein.
Specifications

Chemical Identifiers

123-51-3
88.15
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
CHEBI:15837
CC(C)CCO
C5H12O
PHTQWCKDNZKARW-UHFFFAOYSA-N
31260
3-methylbutan-1-ol

Specifications

123-51-3
15, 7354
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methylbutan-1-ol
31260
Liquid
Pass Test
330nm (0.2 AU max.), 405nm (0.1 AU), 510nm (0.1nm)
400mL
C5H12O
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
CC(C)CCO
88.15
CHEBI:15837
≥99%
Coated Amber Glass/Poison Pack
4.3
Phenol/Chloroform/Isoamyl Alcohol, 125:24:1 Mixture
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

Recommended Storage : 4°C