missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol AR Liquefied, J.T.Baker™

Chemical Identifiers

CAS 108-95-2
Physical Form Liquid
Molecular Formula C6H5OH
Formula Weight 94.11
Chemical Name or Material Phenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK0025212 Avantor Performance
002506
Glass 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK0025500 Avantor Performance
002504
Glass 12 x 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

108-95-2
C6H5OH
Phenol
Liquid
94.11

Specifications

2.5kg
Glass
C6H5OH
Phenol
108-95-2
Liquid
94.11