Please login to your online account to display your discounted pricing

Phenol AR Liquefied, J.T.Baker™


Quantity 2.5kg
Packaging Glass

Manufacturer: avantor performance  002506

 View more versions of this product

Catalog No. MK0025212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 94.11
Quantity 2.5kg
Molecular Formula C6H5OH
Chemical Name or Material Phenol
CAS 108-95-2
Physical Form Liquid
Packaging Glass

Specifications

Chemical Name or Material Phenol
CAS 108-95-2
Physical Form Liquid
Molecular Formula C6H5OH
Formula Weight 94.11