Please login to your online account to display your discounted pricing

Phenol AR Liquefied, J.T.Baker™


Quantity 12 x 500g
Packaging Glass

Manufacturer: avantor performance  002504

 View more versions of this product

Catalog No. MK0025500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 94.11
Physical Form Liquid
Quantity 12 x 500g
Packaging Glass
Molecular Formula C6H5OH
Chemical Name or Material Phenol
CAS 108-95-2

Specifications

Chemical Name or Material Phenol
CAS 108-95-2
Physical Form Liquid
Molecular Formula C6H5OH
Formula Weight 94.11