Please login to your online account to display your discounted pricing

Phenol ACS AR Loose Crystals, J.T.Baker™


Quantity 125g
Packaging Poison Pack

Manufacturer: avantor performance  027303

 View more versions of this product

Catalog No. MK0273125

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 94.11
Physical Form Loose Crystals
Packaging Poison Pack
Chemical Name or Material Phenol
Quantity 125g
Molecular Formula C6H5OH
CAS 108-95-2

Specifications

Chemical Name or Material Phenol
CAS 108-95-2
Purity Grade ACS Grade
Physical Form Loose Crystals
Molecular Formula C6H5OH
Formula Weight 94.11