Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenol, ACS, 99+%, stab.

$48.66 - $76.09

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Name Note Stabilized
Purity Grade ACS Grade
Formula Weight 94.11
MDL Number MFCD00002143
UN Number UN1671
Beilstein 969616
Merck Index 14,7241
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3321322

  SDS
alfa aesar™
3321322
100g Each for $48.66

AA3321336

  SDS
alfa aesar™
3321336
500g Each for $76.09

AA33213A3

  SDS
alfa aesar™
33213A3
2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 94.11
Health Hazard 2 GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Phenol
TSCA TSCA
Name Note Stabilized
Odor Sweet
Purity Grade ACS Grade
Physical Form Crystalline
Assay ≥99%
MDL Number MFCD00002143
CAS 108-95-2
UN Number UN1671
Beilstein 969616
Merck Index 14,7241
Health Hazard 1 Danger
Density 1.071g/mL
Boiling Point 181°C
Melting Point 40° to 43°C
Flash Point 79° (175°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID
RTECS Number SJ3325000
Sensitivity Air Sensitive
Quantity 100g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.