missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 180785000

 View more versions of this product

Catalog No. AC180785000




Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenol
108-95-2
Colorless to White
182.0°C
Authentic
Glass bottle
C6H5OH
500 g
0.05% max.
1.07
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
CHEBI:15882
99+%
Loose Crystals
99+%
39.0°C to 42.0°C
1.0700g/mL
79°C
99% min. (GC)
C6H6O
MFCD00002143
06, 110
01,828; 13,166; 14,88
15, 7354
Solubility in water: 8.4g/L (20°C). Other solubilities: soluble in aq. alkali hydroxides, soluble in alcohol, chloroform, ether, glycerol,, carbon disulfide and fixed oils, 1g/12mL benzene, practically insoluble in petroleum ether
OC1=CC=CC=C1
94.11
3.437 mPa.s (50°C)
94.11
Biochemistry

Chemical Identifiers

108-95-2
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
phenol
C6H6O
MFCD00002143
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
CHEBI:15882
OC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 203-632-7

RTECSNumber : SJ3325000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only