Please login to your online account to display your discounted pricing

Phenol, 99+%, ACS reagent, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Purity Grade ACS Reagent
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C6H6O
Linear Formula C6H5OH
Formula Weight 94.11g/mol
MDL Number MFCD00002143
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC417170025

  SDS
thermo fisher scientific
417170025
Plastic drum 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Clarity Passes Test
Infrared Spectrum Authentic
Packaging Plastic drum
Residue after Evaporation 0.05% max.
Storage Information May become pink on exposure to air
Water 0.5% max. (K.F.)
Density 1.0700g/mL
Color Colorless to white
Melting Point 40.5°C min. ( on dry substance)
Quantity 2.5kg
Chemical Name or Material Phenol
CAS 108-95-2
Purity Grade ACS Reagent
Physical Form Colorless to white crystals or crystalline powder
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C6H6O
Linear Formula C6H5OH
Formula Weight 94.11g/mol
MDL Number MFCD00002143
Beilstein 06, 110
Fieser 01,828; 13,166; 14,88
Merck Index 15, 7354
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number SJ3325000
EINECS Number 203-632-7
TSCA TSCA
Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.