Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenol, 99.5%, unstab.

Molecular biology

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Name Note UnStabilized
Molecular Formula C6H5OH
Formula Weight 94.11
MDL Number MFCD00002143
UN Number UN1671
Beilstein 969616
Merck Index 14,7241
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4452622

  SDS
alfa aesar™
4452622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solid Melt
Formula Weight 94.11
Chemical Name or Material Phenol
TSCA TSCA
Name Note UnStabilized
Odor Sweet
Assay 99.5%
Molecular Formula C6H5OH
MDL Number MFCD00002143
CAS 108-95-2
UN Number UN1671
Beilstein 969616
Merck Index 14,7241
Health Hazard 1 Danger
Density 1.071g/mL
Health Hazard 2 GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Melting Point 40° to 43°C
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID
RTECS Number SJ3325000
Sensitivity Hygroscopic
Quantity 100g
Boiling Point 181°C
Flash Point 79°C (175°F)
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.