Please login to your online account to display your discounted pricing

Phenol, 5% (w/v) Aqueous Solution, AOAC/APHA for Selenium, Ricca Chemical

$59.03 - $98.48

Chemical Identifiers

Chemical Name or Material Phenol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5738-32

  SDS
ricca chemical company
RI573832
Glass 1L Each for $98.48

5738-16

  SDS
ricca chemical company
RI573816
Glass 500mL Each for $59.03
Description & Specifications

Specifications

Quantity 1L
Methods AOAC/APHA for Selenium, AOAC for Disinfectant Testing
Concentration or Composition (by Analyte or Components) 5% (w/v) Aqueous Solution
Chemical Name or Material Phenol
Packaging Glass