missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenetole, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 221495000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenetole | |
103-73-1 | |
Colorless to Yellow | |
169.0°C to 170.0°C | |
Authentic | |
Glass bottle | |
1.5055 to 1.5075 | |
MFCD00009090 | |
06, 140 | |
15, 7343 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and carbon tetrachloride | |
CCOC1=CC=CC=C1 | |
122.17 | |
CHEBI:67129 | |
98+% |
99% | |
-30.0°C | |
0.9600g/mL | |
57°C | |
98% min. (GC) | |
C8H10O | |
C6H5OC2H5 | |
500 mL | |
0.96 | |
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f | |
DLRJIFUOBPOJNS-UHFFFAOYSA-N | |
ethoxybenzene | |
7674 | |
122.17 | |
Liquid |
Chemical Identifiers
103-73-1 | |
122.17 | |
DLRJIFUOBPOJNS-UHFFFAOYSA-N | |
7674 | |
ethoxybenzene |
C8H10O | |
MFCD00009090 | |
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f | |
CHEBI:67129 | |
CCOC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 203-139-7
RTECSNumber : SI7700000
TSCA : TSCA
RUO – Research Use Only