missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenethyl Caffeate 98.0+%, TCI America™
Supplier: TCI America P2088250MG
Specifications
Phenethyl Caffeate | |
129°C | |
C17H16O4 | |
250 mg | |
SWUARLUWKZWEBQ-VQHVLOKHSA-N | |
2-phenylethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate | |
5281787 | |
284.31 | |
Crystalline Powder |
104594-70-9 | |
White-Yellow | |
MFCD00866470 | |
caffeic acid phenethyl ester, phenethyl caffeate, cape, capeee, phenethyl 3-3,4-dihydroxyphenyl acrylate, phenylethyl caffeate, unii-g960r9s5sk, caffeic acid phenylethyl ester, 2-phenylethyl caffeate, caffeic acid 2-phenylethyl ester | |
OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O | |
284.31 | |
CHEBI:8062 | |
≥98.0% (HPLC) |
Chemical Identifiers
104594-70-9 | |
284.31 | |
SWUARLUWKZWEBQ-VQHVLOKHSA-N | |
5281787 | |
2-phenylethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
C17H16O4 | |
MFCD00866470 | |
caffeic acid phenethyl ester, phenethyl caffeate, cape, capeee, phenethyl 3-3,4-dihydroxyphenyl acrylate, phenylethyl caffeate, unii-g960r9s5sk, caffeic acid phenylethyl ester, 2-phenylethyl caffeate, caffeic acid 2-phenylethyl ester | |
CHEBI:8062 | |
OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O |
Safety and Handling
RTECSNumber : UD3334375
TSCA : No
Recommended Storage : Freezer