Please login to your online account to display your discounted pricing

Phenethyl benzoate, +98%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC393582500

 View more versions of this product

Catalog No. AC393582500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 226.28
Physical Form Liquid
Quantity 250g
Color Clear colorless
Density 1.094g/mL
CAS 94-47-3
Molecular Formula C15H14O2
Linear Formula C6H5COOCH2CH2C6H5
Assay Percent Range 98+%
MDL Number MFCD00020681
Packaging Glass bottle
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material Phenethyl benzoate

Specifications

Chemical Name or Material Phenethyl benzoate
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C15H14O2
Linear Formula C6H5COOCH2CH2C6H5
Formula Weight 226.28
MDL Number MFCD00020681