missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenethyl benzoate, +98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393580500

 View more versions of this product

Catalog No. AC393580500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Phenethyl benzoate
94-47-3
Glass bottle
≥98.0%
C15H14O2
C6H5COOCH2CH2C6H5
MFCD00020681
Clear colorless
50g
Authentic
Liquid
98+%
1.5590 to 1.5610 (20°C, 589nm)
226.28
1.094g/mL

Chemical Identifiers

98+%
C6H5COOCH2CH2C6H5
MFCD00020681
≥98.0%
C15H14O2
226.28
Phenethyl benzoate