Please login to your online account to display your discounted pricing

Phenethyl benzoate, +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Phenethyl benzoate
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C15H14O2
Linear Formula C6H5COOCH2CH2C6H5
Formula Weight 226.28
MDL Number MFCD00020681
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393582500

  SDS
thermo fisher scientific
AC393582500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393580500

  SDS
thermo fisher scientific
AC393580500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 226.28
Physical Form Liquid
Quantity 250g
Color Clear colorless
Density 1.094g/mL
CAS 94-47-3
Molecular Formula C15H14O2
Linear Formula C6H5COOCH2CH2C6H5
Assay Percent Range 98+%
MDL Number MFCD00020681
Packaging Glass bottle
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material Phenethyl benzoate