missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenethyl alcohol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 130180010

 View more versions of this product

Catalog No. AC130180010




Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenethyl alcohol
-27.0°C
1.0200g/mL
102°C
98.5% min. (GC)
C8H10O
C6H5CH2CH2OH
1 L
1.02
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
WRMNZCZEMHIOCP-UHFFFAOYSA-N
2-phenylethanol
6054
CHEBI:49000
99%
60-12-8
Colorless
219.0°C
Authentic
Glass bottle
1.5300 to 1.5350
MFCD00002886
06, 478
15, 7338
Solubility in water: 20g/L (20°C). Other solubilities: miscible with alcohol and ether
OCCC1=CC=CC=C1
122.17
7.58 mPa.s (25°C)
122.17
Liquid

Chemical Identifiers

60-12-8
122.17
WRMNZCZEMHIOCP-UHFFFAOYSA-N
6054
OCCC1=CC=CC=C1
C8H10O
MFCD00002886
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
CHEBI:49000
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Warning

EINECSNumber : 200-456-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only