Please login to your online account to display your discounted pricing

Alfa Aesar™ Phencyclone, 97%

$67.58 - $235.51

Chemical Identifiers

Chemical Name or Material Phencyclone
CAS 5660-91-3
Molecular Formula C29H18O
Formula Weight 382.46
MDL Number MFCD00051451
Beilstein 2061817
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0010803

  SDS
alfa aesar™
L0010803
1g Each for $67.58

AAL0010806

  SDS
alfa aesar™
L0010806
5g Each for $235.51
Description & Specifications

Specifications

Formula Weight 382.46
Chemical Name or Material Phencyclone
Molecular Formula C29H18O
Assay 97%
MDL Number MFCD00051451
CAS 5660-91-3
Beilstein 2061817
Melting Point 245° to 249°C
Quantity 1g