Learn More
Phenazine methosulfate, 98+%
CAS: 299-11-6 | C14H14N2O4S | 306.336 g/mol
Supplier: Thermo Scientific Chemicals H5671806
Description
It is used with ascorbic acid to determine nitric oxide reductase activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used with ascorbic acid to determine nitric oxide reductase activity.
Solubility
Soluble in water. (100g/L at20°C)
Notes
Light sensitive. Incompatible with oxidizing agents. Store at -20°C.
Specifications
| Phenazine methosulfate | |
| C14H14N2O4S | |
| 5 g | |
| Light sensitive | |
| Soluble in water. (100g/L at20°C) | |
| C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-] | |
| 306.336 | |
| CHEBI:8055 | |
| ≥98% |
| 299-11-6 | |
| MFCD00011923 | |
| 3898869 | |
| phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate | |
| RXGJTUSBYWCRBK-UHFFFAOYSA-M | |
| 5-methylphenazin-5-ium;methyl sulfate | |
| 9285 | |
| 306.34 |
Chemical Identifiers
| 299-11-6 | |
| 306.336 | |
| RXGJTUSBYWCRBK-UHFFFAOYSA-M | |
| 9285 | |
| 5-methylphenazin-5-ium;methyl sulfate |
| C14H14N2O4S | |
| MFCD00011923 | |
| phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate | |
| CHEBI:8055 | |
| C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-] |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-072-1
RTECSNumber : SG1645000
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only