Please login to your online account to display your discounted pricing

Phenazine methosulfate, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  130160100

 View more versions of this product

Catalog No. AC130160100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 306.33
Physical Form Crystalline Powder
Quantity 10g
Color Dark yellow to brown or khaki
EINECS Number 206-072-1
RTECS Number SG1645000
TSCA TSCA
Merck Index 15, 6182
Synonym N-Methylphenazonium methosulfate
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes serious eye irritation.
UV_Vis 97.5% min. (0.01g/L H2O, 1cm cell, lambda max 387.5nm)
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 1 MUTAGEN
MDL Number MFCD00011923
Name Note 99%
CAS 299-11-6
Assay Percent Range 98%
Chemical Name or Material Phenazine methosulfate
Molecular Formula C13H11N2·CH3O4S

Specifications

Chemical Name or Material Phenazine methosulfate
Name Note 99%
CAS 299-11-6
Assay Percent Range 98%
Molecular Formula C13H11N2·CH3O4S
Formula Weight 306.33
MDL Number MFCD00011923
Synonym N-Methylphenazonium methosulfate
Accessories

MUTAGEN

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents