Learn More
Phenazine, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1577006
Description
Phenazine is the parent substance of many dyestuffs, such as the toluylene red, indulines and safranines. It can be applied in therapeutic uses, used as antipsychotic and antiemetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilityInsoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Phenazine | |
175°C to 177°C | |
MFCD00005023 | |
dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs | |
PCNDJXKNXGMECE-UHFFFAOYSA-N | |
phenazine | |
4757 | |
180.21 |
92-82-0 | |
C12H8N2 | |
5 g | |
Insoluble in water. | |
C1=CC=C2C(=C1)N=C3C=CC=CC3=N2 | |
180.21 | |
CHEBI:36674 | |
≥99% |
Chemical Identifiers
92-82-0 | |
180.21 | |
PCNDJXKNXGMECE-UHFFFAOYSA-N | |
4757 | |
phenazine |
C12H8N2 | |
MFCD00005023 | |
dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs | |
CHEBI:36674 | |
C1=CC=C2C(=C1)N=C3C=CC=CC3=N2 |
Safety and Handling
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H341
EINECSNumber : 202-193-9
RTECSNumber : SG1360000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only