Please login to your online account to display your discounted pricing

Phenazine 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC130151000

 View more versions of this product

Catalog No. AC130151000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.21
Physical Form Crystalline Powder
Assay ≥97.5%
Quantity 100g
Color Yellow to brown
TSCA TSCA
RTECS Number SG1360000
EINECS Number 202-193-9
Melting Point 174° to 177°C
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects.
Packaging Glass bottle
Molecular Formula C12H8N2
CAS 92-82-0
Name Note 98%
Assay Percent Range 98%
Chemical Name or Material Phenazine
MDL Number MFCD00005023

Specifications

Chemical Name or Material Phenazine
Name Note 98%
CAS 92-82-0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H8N2
Formula Weight 180.21
MDL Number MFCD00005023
Accessories

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects.