Please login to your online account to display your discounted pricing

Phenanthrenequinone, 95%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  168791000

 View more versions of this product

Catalog No. AC168791000

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Orange-brownish
Formula Weight 208.22
Physical Form Powder
Assay ≥94%
Quantity 100g
Beilstein 07, 796
Merck Index 15, 7327
Fieser 01,827; 02,317
TSCA TSCA
RTECS Number SF7875000
EINECS Number 201-515-5
Melting Point 203° to 211°C
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Chemical Name or Material Phenanthrenequinone
CAS 84-11-7
MDL Number MFCD00001163
Molecular Formula C14H8O2
Name Note 95%
Assay Percent Range 95%

Specifications

Chemical Name or Material Phenanthrenequinone
Name Note 95%
CAS 84-11-7
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H8O2
Formula Weight 208.22
MDL Number MFCD00001163
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents