Please login to your online account to display your discounted pricing

Phenanthrene 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  130090050

 View more versions of this product

Catalog No. AC130090050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 178.23
Quantity 5g
Color White to gray
Beilstein 05, 667
Merck Index 15, 7326
EINECS Number 201-581-5
TSCA TSCA
Assay 97%
RTECS Number SF7175000
Density 1.063g/mL
Melting Point 99° to 101°C
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Phenanthrene
Name Note 98+%
Assay Percent Range 97%
Molecular Formula C14H10
MDL Number MFCD00001168
CAS 85-01-8

Specifications

Chemical Name or Material Phenanthrene
Name Note 98+%
CAS 85-01-8
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H10
Formula Weight 178.23
MDL Number MFCD00001168
Accessories

GHS Signal Word: Warning

GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

SDS & Safety Documents