missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenanthrene-9-boronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5254606

 View more versions of this product

Catalog No. AAH5254606



Description

Description

Phenanthrene-9-boronic acid as a reactant is involved in the synthesis of large polycyclic aromatic hydrocarbons (PAHs) and electron rich benzo[g,h,i]perylenes. It is also used for preparation of FLAP protein inhibitors as anti-inflammatory agents and aminoisoquinolinones as PARP-2 selective inhibitors. As a reactant it is involved in homocoupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene-9-boronic acid as a reactant is involved in the synthesis of large polycyclic aromatic hydrocarbons (PAHs) and electron rich benzo[g,h,i]perylenes. It is also used for preparation of FLAP protein inhibitors as anti-inflammatory agents and aminoisoquinolinones as PARP-2 selective inhibitors. As a reactant it is involved in homocoupling reactions.

Solubility
Soluble in DMSO.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Phenanthrene-9-boronic acid
254°C to 256°C
MFCD00143524
9-phenanthreneboronic acid, 9-phenanthracenylboronic acid, phenanthrene-9-boronic acid, 9-phenanthrenylboronic acid, 9-phenanthrene boronic acid, 9-phenanthreneboronicacid, 9-phenanthrylboronic acid, boronic acid, 9-phenanthrenyl, phenanthracene-9-boronic acid, phenanthreneboronicacid
JCDAUYWOHOLVMH-UHFFFAOYSA-N
phenanthren-9-ylboronic acid
11775704
97%
68572-87-2
C14H11BO2
5 g
Soluble in DMSO.
OB(O)C1=CC2=CC=CC=C2C2=CC=CC=C12
222.05
222.05

Chemical Identifiers

68572-87-2
222.05
JCDAUYWOHOLVMH-UHFFFAOYSA-N
11775704
OB(O)C1=CC2=CC=CC=C2C2=CC=CC=C12
C14H11BO2
MFCD00143524
9-phenanthreneboronic acid, 9-phenanthracenylboronic acid, phenanthrene-9-boronic acid, 9-phenanthrenylboronic acid, 9-phenanthrene boronic acid, 9-phenanthreneboronicacid, 9-phenanthrylboronic acid, boronic acid, 9-phenanthrenyl, phenanthracene-9-boronic acid, phenanthreneboronicacid
phenanthren-9-ylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only