missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenanthrene-4,5-dicarboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H54939MD

 View more versions of this product

Catalog No. AAH54939MD



Description

Description

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Specifications

Specifications

Phenanthrene-4,5-dicarboxylic acid
250mg
C16H10O4
4,5-phenanthrenedicarboxylic acid
C1=CC2=C(C(=C1)C(=O)O)C3=C(C=CC=C3C(=O)O)C=C2
266.252
CHEBI:81603
97%
244°C to 246°C
5462-82-8
MFCD11110581
RAGXPRCIICFJIK-UHFFFAOYSA-N
phenanthrene-4,5-dicarboxylic acid
95075
266.25

Chemical Identifiers

5462-82-8
266.252
RAGXPRCIICFJIK-UHFFFAOYSA-N
95075
phenanthrene-4,5-dicarboxylic acid
C16H10O4
MFCD11110581
4,5-phenanthrenedicarboxylic acid
CHEBI:81603
C1=CC2=C(C(=C1)C(=O)O)C3=C(C=CC=C3C(=O)O)C=C2
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents