Please login to your online account to display your discounted pricing

Phenamil, Tocris

Inhibits TRPP3-mediated currents; also inhibits epithelial Na+ channels

$100.81 - $427.46

Chemical Identifiers

Chemical Name or Material Phenamil
Assay Percent Range >99%
Molecular Formula C12H12ClN7O·CH3SO3H
Formula Weight 401.83
Synonym 3, 5-Diamino-6-chloro-N-[imino(phenylamino)methyl]pyrazinecarboxamide methanesulfonate salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

337950

 
r & d systems
337950
50mg Each for $427.46

337910

 
r & d systems
337910
10mg Each for $100.81
Description & Specifications

Specifications

Formula Weight 401.83
Quantity 50mg
CAS 1161-94-0
Synonym 3, 5-Diamino-6-chloro-N-[imino(phenylamino)methyl]pyrazinecarboxamide methanesulfonate salt
Molecular Formula C12H12ClN7O·CH3SO3H
Chemical Name or Material Phenamil
Assay Percent Range >99%
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C