Please login to your online account to display your discounted pricing

Phenacyltriphenylphosphonium bromide, 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  165940250

 View more versions of this product

Catalog No. AC165940250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Amorphous Powder
Formula Weight 461.33
Quantity 25g
Color Light beige
EINECS Number 227-945-3
RTECS Number TA2400000
Melting Point 263° to 268°C
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC
Chemical Name or Material Phenacyltriphenylphosphonium bromide
Name Note 96%
Packaging Glass bottle
Assay Percent Range 96%
Molecular Formula C26 H22 Br O P
Linear Formula C6H5COCH2P(C6H5)3Br
MDL Number MFCD00011920
CAS 6048-29-9
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Phenacyltriphenylphosphonium bromide
Name Note 96%
CAS 6048-29-9
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C26 H22 Br O P
Linear Formula C6H5COCH2P(C6H5)3Br
Formula Weight 461.33
MDL Number MFCD00011920
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents