Please login to your online account to display your discounted pricing

Phenacyltriphenylphosphonium bromide, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Phenacyltriphenylphosphonium bromide
Name Note 96%
CAS 6048-29-9
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C26 H22 Br O P
Linear Formula C6H5COCH2P(C6H5)3Br
Formula Weight 461.33
MDL Number MFCD00011920
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC165940250

  SDS
thermo fisher scientific
165940250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Formula Weight 461.33
Physical Form Amorphous Powder
Quantity 25g
Color Light beige
RTECS Number TA2400000
EINECS Number 227-945-3
Melting Point 263° to 268°C
Health Hazard 1 HYGROSCOPIC
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Assay Percent Range 96%
Chemical Name or Material Phenacyltriphenylphosphonium bromide
Molecular Formula C26 H22 Br O P
MDL Number MFCD00011920
Name Note 96%
Linear Formula C6H5COCH2P(C6H5)3Br
CAS 6048-29-9

For C-C Bond Formation (Olefination)

Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.