Please login to your online account to display your discounted pricing

PHCCC, Tocris


Potent group I mGlu antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS102750

 View more versions of this product

Catalog No. 102750

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 294.31
Quantity 50mg
Molecular Formula C17H14N2O3
CAS 177610-87-6
Chemical Name or Material PHCCC
Assay Percent Range >98%
Synonym N-Phenyl-7-(hydroxyimino)cyclopropa[b]chromen-1a-carboxamide
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material PHCCC
Assay Percent Range >98%
Molecular Formula C17H14N2O3
Formula Weight 294.31
Synonym N-Phenyl-7-(hydroxyimino)cyclopropa[b]chromen-1a-carboxamide
Accessories

Store at Room Temperature