Please login to your online account to display your discounted pricing

Phaclofen, Tocris

Weak, selective GABAB antagonist

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Phaclofen
Assay Percent Range >97%
Molecular Formula C9H13ClNO3P
Formula Weight 249.63
Synonym (RS)-3-Amino-2-(4-chlorophenyl)propylphosphonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

017810

 
r & d systems
RDS017810
10mg Each for $164.61

017850

 
r & d systems
RDS017850
50mg Each for $682.67
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 249.63
Assay Percent Range >97%
Chemical Name or Material Phaclofen
Synonym (RS)-3-Amino-2-(4-chlorophenyl)propylphosphonic acid
Molecular Formula C9H13ClNO3P
CAS 114012-12-3
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature