Oakton™ pH Singles Calibration Solutions in 20mL Pouches
Economical pouches are accurate and convenient.
Manufacturer:  Fisher Science Education WD3565301EMD
 View more versions of this product
Catalog No. S66771
Description
Description
The pH Singles are ideal for field calibration—just select the pouches needed and carry them to your test site for on-the-spot calibration and verification. No bottles to carry or clean. Pouches are air-tight for maximum freshness and unquestionable reliability. Packets are labeled with temperature versus pH reference tables. Sold in packs of twenty 20mL pouches.
Certifications
NIST traceable, All solutions are certified traceable to NIST™ standards.
Specifications
Specifications
Education
4.01
20mL Pouches
Chemical Identifiers
0.0
icon-pdf.png
SDS Documents