pH Singles Calibration Solutions in 20mL Pouches
$57.40 - $57.75
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S66771 Fisher Science Education
WD3565301EMD
Pack of 20 Each for $57.75
S66772 Fisher Science Education
WD35653-02E
Pack of 20 Each for $57.75
S66773 Fisher Science Education
WD3565303EMD
Pack of 20 Each for $57.75
S66774 Fisher Science Education
WD35653-04E
Pack of 20 Each for $57.40
Specifications
Description
The pH Singles are ideal for field calibration—just select the pouches needed and carry them to your test site for on-the-spot calibration and verification. No bottles to carry or clean. Pouches are air-tight for maximum freshness and unquestionable reliability. Packets are labeled with temperature versus pH reference tables. Sold in packs of twenty 20mL pouches.
Certifications
NIST traceable, All solutions are certified traceable to NIST™ standards.