Please login to your online account to display your discounted pricing

PG-9 maleate, Tocris


Presynaptic cholinergic modulator

Quantity 50mg

Manufacturer: r & d systems  RDS075050

 View more versions of this product

Catalog No. 075050

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 468.34
Chemical Name or Material PG-9 maleate
Assay Percent Range >98%
Molecular Formula C17H22BrNO2·C4H4O4
CAS 155649-00-6
Synonym Tropanyl 2-(4-bromophenyl)propionate maleate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material PG-9 maleate
Assay Percent Range >98%
Molecular Formula C17H22BrNO2·C4H4O4
Formula Weight 468.34
Synonym Tropanyl 2-(4-bromophenyl)propionate maleate
Accessories

Store at Room Temperature