Please login to your online account to display your discounted pricing

PG-9 maleate, Tocris

Presynaptic cholinergic modulator

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material PG-9 maleate
Assay Percent Range >98%
Molecular Formula C17H22BrNO2·C4H4O4
Formula Weight 468.34
Synonym Tropanyl 2-(4-bromophenyl)propionate maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

075050

 
r & d systems
RDS075050
50mg Each for $682.67

075010

 
r & d systems
RDS075010
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 468.34
Synonym Tropanyl 2-(4-bromophenyl)propionate maleate
Molecular Formula C17H22BrNO2·C4H4O4
Assay Percent Range >98%
Chemical Name or Material PG-9 maleate
Recommended Storage Store at Room Temperature
CAS 155649-00-6
Safety and Handling

Store at Room Temperature